4-(Trifluoromethylthio)phenylacetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

005535-1G 005535-5G 005535-25G
101789-092EA 47.69 USD
101789-092 101789-094 101789-096
4-(Trifluoromethylthio)phenylacetic acid ≥97%
4-(Trifluoromethylthio)phenylacetic acid

Hazard: Irritant

Formula: C₉H₇F₃O₂S
MW: 236.208 g/mol
MDL Number: MFCD00236351
CAS Number: 243977-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR