3-(Trifluoromethyl)phenyl acetate ≥98%

Supplier: Matrix Scientific

005447-5G 005447-25G
101788-970EA 100 USD
101788-970 101788-972
3-(Trifluoromethyl)phenyl acetate ≥98%
3-(Trifluoromethyl)phenyl acetate

Hazard: Irritant

Formula: C₉H₇F₃O₂
MDL Number: MFCD01861206
CAS Number: 78950-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR