8-(Trifluoromethyl)-4-quinolinol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-8-(trifluoromethyl)quinoline

005412-1G 005412-5G
101788-896EA 36.92 USD
101788-896 101788-898
8-(Trifluoromethyl)-4-quinolinol ≥97%
8-(Trifluoromethyl)-4-quinolinol

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 170-174°C
MDL Number: MFCD00134577
CAS Number: 23779-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR