2-Hydrazinyl-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

005408-1G 005408-5G
101788-882EA 33.85 USD
101788-882 101788-884
2-Hydrazinyl-5-(trifluoromethyl)pyridine ≥97%
2-Hydrazinyl-5-(trifluoromethyl)pyridine

Hazard: Irritant/Refrigerate

Formula: C₆H₆F₃N₃
MW: 177.13 g/mol
MDL Number: MFCD00236724
CAS Number: 89570-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR