4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)cyclohexanol, 4-(Trifluoromethyl)cyclohexan-1-ol

005400-1G 005400-5G
101788-854EA 72.31 USD
101788-854 101788-856
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) ≥97%
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)

(mixture of cis/trans isomers) Purity: 97% CAS: 30129-18-1 MDL Number: MFCD00102144 Molecular Formula: C7H11F3O Molecular Weight: 168,16 Boiling Point: 70-75°C/15mm Hazard: Irritant

Formula: C₇H₁₁F₃O
MW: 168.16 g/mol
Boiling Pt: 70-75°C/15mm
MDL Number: MFCD00102144
CAS Number: 30129-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR