2-(Trifluoromethyl)cyclohexanol cis- and trans-mixture, Technical Grade

Supplier: Matrix Scientific

005399-1G 005399-5G
101788-850EA 112.31 USD
101788-850 101788-852
2-(Trifluoromethyl)cyclohexanol cis- and trans-mixture, Technical Grade
2-(Trifluoromethyl)cyclohexanol

Hazard: Irritant

Formula: C₇H₁₁F₃O
MDL Number: MFCD00102142
CAS Number: 104282-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR