6-(Trifluoromethoxy)-4-quinolinol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-6-(trifluoromethoxy)quinoline, 6-(Trifluoromethoxy)quinolin-4-ol

005332-250MG 005332-1G
101788-678EA 80 USD
101788-678 101788-680
6-(Trifluoromethoxy)-4-quinolinol ≥97%
6-(Trifluoromethoxy)-4-quinolinol

Hazard: Irritant

Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
MDL Number: MFCD00219858
CAS Number: 175203-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR