3-Methyl-1H-indazole-6-boronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methyl-1H-indazole-6-boronic Acid, 3-Methylindazole-6-boronic acid
070424-500MG
070424-1G
070424-5G
101944-686EA
1106.15
USD
101944-686
101944-688
101944-690
3-Methyl-1H-indazole-6-boronic acid ≥95%
3-Methyl-1H-indazole-6-boronic acid
Hazard: Irritant
Formula:
C₈H₉BN₂O₂ MW: 175.9800 g/mol |
MDL Number:
MFCD11044596 CAS Number: 1245816-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...