3-Methyl-1H-indazole-5-boronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-1H-indazol-5-yl-5-boronic acid

070422-500MG 070422-1G 070422-5G
101944-676EA 930.77 USD
101944-676 101944-678 101944-680
3-Methyl-1H-indazole-5-boronic acid ≥95%
3-Methyl-1H-indazole-5-boronic acid

Hazard: Irritant

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
MDL Number: MFCD12028643
CAS Number: 1245816-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR