(S)-tert-Butyl(pyrrolidin-3-ylmethyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl (pyrrolidin-3-ylmethyl)carbamate

070421-1G 070421-5G
101944-672EA 346.15 USD
101944-672 101944-674
(S)-tert-Butyl(pyrrolidin-3-ylmethyl)carbamate ≥97%
(S)-tert-Butyl(pyrrolidin-3-ylmethyl)carbamate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD06796616
CAS Number: 173340-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR