7-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidine
Supplier: Matrix Scientific
070420-1G
070420-5G
101944-668EA
1221.54
USD
101944-668
101944-670
7-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidine
7-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidine
Hazard: Irritant
Formula:
C₈H₈ClN₃ |
MDL Number:
MFCD11043774 CAS Number: 136549-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...