2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
Supplier: Matrix Scientific
070419-1G
070419-5G
070419-25G
101944-662EA
263.08
USD
101944-662
101944-664
101944-666
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
Hazard: Irritant
Formula:
C₈H₉N₃O MW: 163.18 g/mol |
MDL Number:
MFCD00210153 CAS Number: 98488-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...