2-Methyl-4-(tributylstannyl)pyridine ≥95%

Supplier: Matrix Scientific

070418-1G 070418-5G
101944-658EA 553.85 USD
101944-658 101944-660
2-Methyl-4-(tributylstannyl)pyridine ≥95%
2-Methyl-4-(tributylstannyl)pyridine

Hazard: Irritant/Refrigerate

Formula: C₁₈H₃₃NSn
MDL Number: MFCD16039584
CAS Number: 134914-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR