6-Bromo-1H-indazole-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
070415-1G
070415-5G
101944-644EA
930.77
USD
101944-644
101944-646
6-Bromo-1H-indazole-3-carboxylic acid ≥97%
6-Bromo-1H-indazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅BrN₂O₂ |
MDL Number:
MFCD05663977 CAS Number: 660823-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...