4-(Trimethylsilyl)-3-butyn-1-amine ≥94%

Supplier: Matrix Scientific

070411-1G 070411-5G
101944-630EA 930.77 USD
101944-630 101944-632
4-(Trimethylsilyl)-3-butyn-1-amine ≥94%
4-(Trimethylsilyl)-3-butyn-1-amine

Hazard: Irritant

Formula: C₇H₁₅NSi
MW: 141.29 g/mol
MDL Number: MFCD14702527
CAS Number: 183208-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR