6-Bromo-1-chloroisoquinoline ≥97%
Supplier: Matrix Scientific
070409-1G
070409-5G
101944-622EA
136.92
USD
101944-622
101944-624
6-Bromo-1-chloroisoquinoline ≥97%
6-Bromo-1-chloroisoquinoline
Hazard: Irritant
Formula:
C₉H₅BrClN |
MDL Number:
MFCD06738661 CAS Number: 205055-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...