1,3,6-Trichloroisoquinoline ≥97%

Supplier: Matrix Scientific

070408-1G 070408-5G
101944-618EA 407.69 USD
101944-618 101944-620
1,3,6-Trichloroisoquinoline ≥97%
1,3,6-Trichloroisoquinoline

Hazard: Irritant

Formula: C₉H₄Cl₃N
MDL Number: MFCD09907620
CAS Number: 1053658-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR