6-Bromo-1,3-dichloroisoquinoline ≥97%

Supplier: Matrix Scientific

070406-1G 070406-5G
101944-610EA 233.85 USD
101944-610 101944-612
6-Bromo-1,3-dichloroisoquinoline ≥97%
6-Bromo-1,3-dichloroisoquinoline

Hazard: Irritant

Formula: C₉H₄BrCl₂N
MW: 276.9448 g/mol
MDL Number: MFCD09258639
CAS Number: 552331-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR