3-Chloro-1-methoxyisoquinoline ≥97%

Supplier: Matrix Scientific

070405-1G 070405-5G
101944-606EA 330.77 USD
101944-606 101944-608
3-Chloro-1-methoxyisoquinoline ≥97%
3-Chloro-1-methoxyisoquinoline

Hazard: Irritant

Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
MDL Number: MFCD00160677
CAS Number: 24649-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR