4-(Phenylthio)-2(5H)-furanone ≥96%

Supplier: Matrix Scientific

070404-1G 070404-5G
101944-602EA 1106.15 USD
101944-602 101944-604
4-(Phenylthio)-2(5H)-furanone ≥96%
4-(Phenylthio)-2(5H)-furanone

Hazard: Irritant

Formula: C₁₀H₈O₂S
MDL Number: MFCD14702525
CAS Number: 57061-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR