3-(Phenylthio)-2(5H)-furanone ≥96%

Supplier: Matrix Scientific

070403-1G 070403-5G
101944-598EA 930.77 USD
101944-598 101944-600
3-(Phenylthio)-2(5H)-furanone ≥96%
3-(Phenylthio)-2(5H)-furanone

Hazard: Irritant

Formula: C₁₀H₈O₂S
MDL Number: MFCD14702524
CAS Number: 42435-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR