6-Chloroimidazo[1,2-b]pyridazinemethanol ≥97%

Supplier: Matrix Scientific

070400-1G 070400-5G
101944-586EA 1396.92 USD
101944-586 101944-588
6-Chloroimidazo[1,2-b]pyridazinemethanol ≥97%
6-Chloroimidazo[1,2-b]pyridazinemethanol

Hazard: Irritant

Formula: C₇H₆ClN₃O
MW: 183.6 g/mol
MDL Number: MFCD12406086
CAS Number: 1184916-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR