5-Bromo-3-hydroxy-1H-indole-2-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

070399-1G 070399-5G
101944-582EA 938.46 USD
101944-582 101944-584
5-Bromo-3-hydroxy-1H-indole-2-carboxylic acid ethyl ester ≥95%
5-Bromo-3-hydroxy-1H-indole-2-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₃
MDL Number: MFCD06659772
CAS Number: 153501-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR