3-(Trimethylsilyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: (3-(Trimethylsilyl)phenyl)boronic acid

070396-1G 070396-5G
101944-566EA 263.08 USD
101944-566 101944-568
3-(Trimethylsilyl)phenylboronic acid
3-(Trimethylsilyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₅BO₂Si
MW: 194.1100 g/mol
MDL Number: MFCD03093884
CAS Number: 177171-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR