N-(Methoxymethyl)-2-methyl-N-[(trimethylsilyl)-methyl]-2-propanamine ≥97%

Supplier: Matrix Scientific

070395-1G 070395-5G 070395-25G
101944-560EA 263.08 USD
101944-560 101944-562 101944-564
N-(Methoxymethyl)-2-methyl-N-[(trimethylsilyl)-methyl]-2-propanamine ≥97%
N-(Methoxymethyl)-2-methyl-N-[(trimethylsilyl)-methyl]-2-propanamine

Hazard: Irritant

Formula: C₁₁H₂₇NOSi
MDL Number: MFCD14702523
CAS Number: 455954-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR