6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

070392-1G 070392-5G
101944-546EA 1629.23 USD
101944-546 101944-548
6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid ≥97%
6-Chloropyrazolo[1,5-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD11518928
CAS Number: 876379-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR