cis-1-(Tributylstannyl)-1-propene ≥90%

Supplier: Matrix Scientific

070391-1G 070391-5G
101944-542EA 349.23 USD
101944-542 101944-544
cis-1-(Tributylstannyl)-1-propene ≥90%
cis-1-(Tributylstannyl)-1-propene

Hazard: Irritant

Formula: C₁₅H₃₂Sn
CAS Number: 66680-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR