1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers) ≥96%

Supplier: Matrix Scientific

Synonyms: Tri-n-butyl(1-propenyl)tin

070390-1G 070390-5G
101944-538EA 183.08 USD
101944-538 101944-540
1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers) ≥96%
1-(Tributylstannyl)-1-propene (mixture of cis and trans isomers)

Hazard: Irritant/Refrigerate

Formula: C₁₅H₃₂Sn
MW: 331.13
MDL Number: MFCD03425868
CAS Number: 105494-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR