(E)-3-Methoxy-2-butenoic acid methyl ester ≥96%

Supplier: Matrix Scientific

070389-1G 070389-5G
101944-534EA 524.62 USD
101944-534 101944-536
(E)-3-Methoxy-2-butenoic acid methyl ester ≥96%
(E)-3-Methoxy-2-butenoic acid methyl ester

Hazard: Irritant

Formula: C₆H₁₀O₃
MDL Number: MFCD09838566
CAS Number: 4525-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR