(E)-3-Methoxy-2-butenoic acid ≥96%

Supplier: Matrix Scientific

070388-1G 070388-5G
101944-530EA 923.08 USD
101944-530 101944-532
(E)-3-Methoxy-2-butenoic acid ≥96%
(E)-3-Methoxy-2-butenoic acid

Hazard: Irritant/Store in Freezer

Formula: C₅H₈O₃
MDL Number: MFCD14525462
CAS Number: 156948-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR