2-(Tributylstannyl)oxazole ≥95%

Supplier: Matrix Scientific

070387-1G 070387-5G
101944-526EA 461.54 USD
101944-526 101944-528
2-(Tributylstannyl)oxazole ≥95%
2-(Tributylstannyl)oxazole

Hazard: Toxic

Formula: C₁₅H₂₉NOSn
Boiling Pt: 108-110°C/0.2mm
Density: 1.71
MDL Number: MFCD06798076
CAS Number: 145214-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR