2-(Tributylstannyl)furan ≥97%

Supplier: Matrix Scientific

070385-1G 070385-5G 070385-25G
101944-516EA 76.92 USD
101944-516 101944-518 101944-520
2-(Tributylstannyl)furan ≥97%
2-(Tributylstannyl)furan

Hazard: Toxic

Formula: C₁₆H₃₀OSn
Boiling Pt: 90°C/0.1mm
Density: 1.134
MDL Number: MFCD00192512
CAS Number: 118486-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR