(Phenylethynyl)tributyltin ≥95%

Supplier: Matrix Scientific

070384-1G 070384-5G 070384-25G
101944-510EA 92.31 USD
101944-510 101944-512 101944-514
(Phenylethynyl)tributyltin ≥95%
(Phenylethynyl)tributyltin

Hazard: Irritant

Formula: C₂₀H₃₂Sn
MDL Number: MFCD00075186
CAS Number: 3757-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR