5-Iodo-1,3-dimethyluracil ≥98%

Supplier: Matrix Scientific

070381-1G 070381-5G 070381-25G
101944-492EA 175.38 USD
101944-492 101944-494 101944-496
5-Iodo-1,3-dimethyluracil ≥98%
5-Iodo-1,3-dimethyluracil

Hazard: Irritant

Formula: C₆H₇IN₂O₂
MW: 266.04 g/mol
MDL Number: MFCD00192228
CAS Number: 40738-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR