1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole ≥96%
Supplier: Matrix Scientific
070377-1G
070377-5G
101944-472EA
1135.38
USD
101944-472
101944-474
1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole ≥96%
1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₁₇H₃₁F₃N₂Sn |
MDL Number:
MFCD06797466 CAS Number: 191606-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...