5-Chloro-2-(methylthio)-4-(tributylstannyl)pyrimidine ≥95%

Supplier: Matrix Scientific

070376-1G 070376-5G
101944-468EA 1396.92 USD
101944-468 101944-470
5-Chloro-2-(methylthio)-4-(tributylstannyl)pyrimidine ≥95%
5-Chloro-2-(methylthio)-4-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₇H₃₁ClN₂SSn
MDL Number: MFCD01318962
CAS Number: 123061-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR