1H-Indazole-5-carbaldehyde ≥97%
Supplier: Matrix Scientific
070374-1G
070374-5G
101944-460EA
175.38
USD
101944-460
101944-462
1H-Indazole-5-carbaldehyde ≥97%
1H-Indazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₈H₆N₂O MW: 146.1500 g/mol |
MDL Number:
MFCD06738280 CAS Number: 253801-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...