7-Isoquinolineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Isoquinolin-7-ylboronic acid

070371-1G 070371-5G 070371-25G
101944-444EA 638.46 USD
101944-444 101944-446 101944-448
7-Isoquinolineboronic acid ≥95%
7-Isoquinolineboronic acid

Hazard: Irritant

Formula: C₉H₈BNO₂
MW: 172.9763 g/mol
MDL Number: MFCD08234617
CAS Number: 1092790-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR