4-(Tributylstannyl)-5H-furan-2-one ≥95%

Supplier: Matrix Scientific

070370-1G 070370-5G
101944-438EA 1403.08 USD
101944-438 101944-440
4-(Tributylstannyl)-5H-furan-2-one ≥95%
4-(Tributylstannyl)-5H-furan-2-one

Hazard: Irritant

Formula: C₁₆H₃₀O₂Sn
MDL Number: MFCD01319088
CAS Number: 145439-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR