3-(Trifluoromethyl)-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%
Supplier: Matrix Scientific
070369-250MG
070369-500MG
070369-1G
101944-432EA
815.38
USD
101944-432
101944-434
101944-436
3-(Trifluoromethyl)-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride ≥95%
3-(Trifluoromethyl)-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₀ClF₃N₂ |
MDL Number:
MFCD13151972 CAS Number: 741736-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...