2-(2-(4-Chlorophenyl)thiazol-4-yl)ethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

070367-1G 070367-5G
101944-424EA 1396.92 USD
101944-424 101944-426
2-(2-(4-Chlorophenyl)thiazol-4-yl)ethanamine hydrochloride ≥95%
2-(2-(4-Chlorophenyl)thiazol-4-yl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₂Cl₂N₂S
MDL Number: MFCD12963845
CAS Number: 1187830-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR