3-Nitro-1H-pyrrolo[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

070366-1G 070366-5G
101944-420EA 841.54 USD
101944-420 101944-422
3-Nitro-1H-pyrrolo[3,2-c]pyridine ≥95%
3-Nitro-1H-pyrrolo[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₅N₃O₂
MDL Number: MFCD08690125
CAS Number: 23612-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR