Methyl-2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate ≥95%

Supplier: Matrix Scientific

070363-1G 070363-5G
101944-412EA 1338.46 USD
101944-412 101944-414
Methyl-2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate ≥95%
Methyl-2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate

Hazard: Irritant

Formula: C₁₆H₁₆ClNO₂S
MDL Number: MFCD12963847
CAS Number: 144457-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR