Ethyl-2-(2-(4-chlorophenyl)thiazol-4-yl)acetate ≥95%

Supplier: Matrix Scientific

070362-1G 070362-5G
101944-408EA 1396.92 USD
101944-408 101944-410
Ethyl-2-(2-(4-chlorophenyl)thiazol-4-yl)acetate ≥95%
Ethyl-2-(2-(4-chlorophenyl)thiazol-4-yl)acetate

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₂S
MDL Number: MFCD01765002
CAS Number: 20287-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR