Ethyl-5-fluoro-1H-indole-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-fluoro-1H-indole-2-carboxylate, Ethyl 5-Fluoroindole-2-carboxylate, 5-Fluoroindole-2-carboxylic acid ethyl ester
070360-1G
070360-5G
101944-404EA
175.38
USD
101944-404
101944-406
Ethyl-5-fluoro-1H-indole-2-carboxylate ≥95%
Ethyl-5-fluoro-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀FNO₂ MW: 207.21 g/mol |
MDL Number:
MFCD00152076 CAS Number: 348-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...