4,6-Dichloro-7-azaindole ≥96%
Supplier: Matrix Scientific
Synonyms:
4,6-Dichloro-1H-pyrrolo[2,3-b]pyridine
070358-1G
070358-5G
101944-396EA
246.15
USD
101944-396
101944-398
4,6-Dichloro-7-azaindole ≥96%
4,6-Dichloro-7-azaindole
Hazard: Irritant
Formula:
C₇H₄Cl₂N₂ MW: 187.0300 g/mol |
MDL Number:
MFCD09880157 CAS Number: 5912-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...