5-Bromo-3-iodo-4,7-diazaindole

Supplier: Matrix Scientific

070357-1G 070357-5G
101944-392EA 2269.23 USD
101944-392 101944-394
5-Bromo-3-iodo-4,7-diazaindole
5-Bromo-3-iodo-4,7-diazaindole

Hazard: Irritant

Formula: C₆H₃BrIN₃
MW: 323.9200 g/mol
MDL Number: MFCD09878696
CAS Number: 875781-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR