5-Chloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

070355-1G 070355-5G
101944-384EA 1230.77 USD
101944-384 101944-386
5-Chloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one ≥95%
5-Chloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₇H₅ClN₂O
MDL Number: MFCD08062555
CAS Number: 136888-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR