S-Methylisothiosemicarbazide hydroiodide ≥96%

Supplier: Matrix Scientific

070354-1G 070354-5G
101944-380EA 29.23 USD
101944-380 101944-382
S-Methylisothiosemicarbazide hydroiodide ≥96%
S-Methylisothiosemicarbazide hydroiodide

Hazard: Irritant

Formula: C₂H₈IN₃S
MDL Number: MFCD00035462
CAS Number: 35600-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR