3H-Imidazo[4,5-c]pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Aza-1H-benzoimidazole, 5-Azabenzimidazole

070353-1G 070353-5G
101944-376EA 58.46 USD
101944-376 101944-378
3H-Imidazo[4,5-c]pyridine ≥97%
3H-Imidazo[4,5-c]pyridine

Hazard: Irritant

Formula: C₆H₅N₃
Melting Pt: 168…172 °C
MDL Number: MFCD00051808
CAS Number: 272-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR